siRNA Details
| Gene Symbol: Enpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057104.2 | NP_476445.2 | 19 | 2175 | ||
| GenBank mRNA | BC081747.1 | - | 19 | 2135 | 7115236 | |
| D28560.1 | - | 21 | 2244 | |||
| DQ131564.1 | - | 19 | 2175 | |||
| FQ233297.1 | - | 5 | 491 | |||

Enpp2
Genomic Map