siRNA Details
| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052980.2 | NP_443212.1 | 2 | 318 | ||
| GenBank mRNA | AF151377.1 | - | 2 | 291 | ||
| BC091138.1 | - | 2 | 319 | |||
| BC107449.1 | - | 1 | 80 | |||
| CO575646.1 | - | 2 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr1i2| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052980.2 | NP_443212.1 | 2 | 318 | ||
| GenBank mRNA | AF151377.1 | - | 2 | 291 | ||
| BC091138.1 | - | 2 | 319 | |||
| BC107449.1 | - | 1 | 80 | |||
| CO575646.1 | - | 2 | 318 | |||