siRNA Details
| Gene Symbol: Braf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236357.2 | XP_006236419.2 | 8 | 1104 | ||
| XM_006236358.2 | XP_006236420.2 | 8 | 1104 | |||
| XM_006236359.2 | XP_006236421.2 | 8 | 1104 | |||
| XM_231692.8 | XP_231692.4 | 9 | 1110 | |||

Braf
Genomic Map