siRNA Details
| Gene Symbol: Clec4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053753.1 | NP_446205.1 | 4 | 657 | ||
| GenBank mRNA | BC098661.1 | - | 4 | 688 | 7384948 | |
| J03734.1 | - | 4 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4f| Gene Symbol: Clec4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053753.1 | NP_446205.1 | 4 | 657 | ||
| GenBank mRNA | BC098661.1 | - | 4 | 688 | 7384948 | |
| J03734.1 | - | 4 | 648 | |||