siRNA Details
| Gene Symbol: Zp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053762.1 | NP_446214.1 | 6 | 935 | ||
| GenBank mRNA | BC127488.1 | - | 6 | 935 | 7308805 | |
| D78482.1 | - | 6 | 935 | |||
| Y10823.1 | - | 6 | 928 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zp3| Gene Symbol: Zp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053762.1 | NP_446214.1 | 6 | 935 | ||
| GenBank mRNA | BC127488.1 | - | 6 | 935 | 7308805 | |
| D78482.1 | - | 6 | 935 | |||
| Y10823.1 | - | 6 | 928 | |||