siRNA Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053787.2 | NP_446239.2 | 15 | 2066 | ||
| XM_006236376.2 | XP_006236438.1 | 13 | 1642 | |||
| GenBank mRNA | AB032019.1 | - | 15 | 2066 | ||
| AF209196.1 | - | 2017 | ||||
| AY762624.1 | - | 14 | 1754 | |||

Trpv5
Genomic Map