siRNA Details
| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053957.1 | NP_446409.1 | 12, 13 | 1507 | ||
| XM_006254495.1 | XP_006254557.1 | 12, 13 | 1507 | |||
| GenBank mRNA | BC097935.1 | - | 9, 10 | 1759 | ||
| Y13413.1 | - | 12, 13 | 1507 | |||

Apbb3
Genomic Map