siRNA Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130406.1 | NP_569090.1 | 10 | 1099 | ||
| GenBank mRNA | AF293459.1 | - | 10 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faf1| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130406.1 | NP_569090.1 | 10 | 1099 | ||
| GenBank mRNA | AF293459.1 | - | 10 | 1099 | ||