This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ERG SIRNA Assays ›
Gene Symbol:
Erg
Gene Name:
v-ets avian erythroblastosis virus E26 oncogene
Gene Aliases:
-
Chromosome Location:
Chr.11: 35645317 - 35749643 on Build Rnor_6.0
UniGene ID:
Rn.47987
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139461

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Erg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133397.2 NP_596888.1 2 258
XM_006248142.2 XP_006248204.1 2 417
XM_006248143.2 XP_006248205.1 2 417
XM_006248145.2 XP_006248207.1 2 416
GenBank mRNA AB031088.1 - 2 258
BC072519.1 - 2 238 7105647
FQ217228.1 - 2 277

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133659, AB073078, AK142379, AB073080, AK050922, and others.

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-592sj:80/100.66.79.144:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline