This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MARF1 SIRNA Assays ›
Gene Symbol:
Marf1 LOC103689943
Gene Name:
meiosis arrest female 1
meiosis arrest female protein 1-like
Gene Aliases:
Lkap
-
Chromosome Location:
Chr.10: 908806 - 953481 on Build Rnor_6.0
Chromosome Location:
Chr.10: 28269 - 72912 on Build Rnor_6.0
UniGene ID:
Rn.27366
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s139535

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Marf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133421.2 NP_596912.1 4 1115
XM_003750779.3 XP_003750827.1 4 2162
XM_006245854.2 XP_006245916.1 4 1089
XM_006245855.2 XP_006245917.1 4 941
XM_006245856.2 XP_006245918.1 4 552
XM_008767442.1 XP_008765664.1 3 1596
GenBank mRNA AB012133.1 - 4 1115
Gene Symbol: LOC103689943
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245686.2 XP_006245748.1 4 2163
XM_006245687.2 XP_006245749.1 4 1089
XM_006245688.1 XP_006245750.1 4 941
XM_006245689.2 XP_006245751.1 4 552
XM_008767447.1 XP_008765669.1 4 1089
XM_008767448.1 XP_008765670.1 3 1596

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
female meiotic division
regulation of gene expression
negative regulation of phosphatase activity
oogenesis
nucleotide binding
RNA binding
nucleic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7svs5:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline