siRNA Details
| Gene Symbol: Fcer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033924.1 | NP_001029096.1 | 6, 7 | 433 | ||
| NM_133550.2 | NP_598234.2 | 5, 6 | 370 | |||
| XM_008768939.1 | XP_008767161.1 | 5, 6 | 597 | |||
| GenBank mRNA | AF381978.1 | - | 5, 6 | 319 | ||
| BC078749.1 | - | 6, 7 | 433 | 7124300 | ||
| X73579.1 | - | 4, 5 | 253 | |||

Fcer2
Genomic Map