siRNA Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133597.1 | NP_598281.1 | 5 | 1030 | ||
| GenBank mRNA | AY574259.1 | - | 5 | 853 | ||
| J04636.1 | - | 5 | 1030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnb3| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133597.1 | NP_598281.1 | 5 | 1030 | ||
| GenBank mRNA | AY574259.1 | - | 5 | 853 | ||
| J04636.1 | - | 5 | 1030 | |||