siRNA Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134367.1 | NP_599194.1 | 10 | 1182 | ||
| XM_008770813.1 | XP_008769035.1 | 10 | 1365 | |||
| XM_008770815.1 | XP_008769037.1 | 10 | 1799 | |||
| XM_008770816.1 | XP_008769038.1 | 11 | 1176 | |||
| XM_008770817.1 | XP_008769039.1 | 10 | 1713 | |||
| GenBank mRNA | BC079027.1 | - | 10 | 1182 | 7114037 | |
| GQ376198.1 | - | 10 | 1091 | |||

Ppp3cc
Genomic Map