siRNA Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763520.1 | XP_008761742.1 | 15 | 1493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgk3| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763520.1 | XP_008761742.1 | 15 | 1493 | ||