This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PKN2 SIRNA Assays ›
Gene Symbol:
Pkn2
Gene Name:
protein kinase N2
Gene Aliases:
Pak-2, Prkcl2
Chromosome Location:
Chr.2: 248735699 - 248840735 on Build Rnor_6.0
UniGene ID:
Rn.30325
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140607

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pkn2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105755.2 NP_001099225.2 19 2891
XM_008761494.1 XP_008759716.1 18 2602

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
cell adhesion
signal transduction
epithelial cell migration
positive regulation of cytokinesis
apical junction assembly
positive regulation of viral genome replication
positive regulation of mitotic cell cycle
cell division
regulation of cell motility
protein serine/threonine kinase activity
protein kinase C activity
ATP binding
kinase activity
histone deacetylase binding
poly(A) RNA binding
RNA polymerase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nqfhd:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline