siRNA Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113570.1 | NP_001107042.1 | 1, 2 | 364 | ||
| GenBank mRNA | CB616240.1 | - | 1, 2 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf3a| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113570.1 | NP_001107042.1 | 1, 2 | 364 | ||
| GenBank mRNA | CB616240.1 | - | 1, 2 | 372 | ||