siRNA Details
| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145682.1 | NP_663715.1 | 5 | 3448 | ||
| GenBank mRNA | AB055759.1 | - | 5 | 3448 | ||
| D87257.1 | - | 2 | 2869 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Filip1| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145682.1 | NP_663715.1 | 5 | 3448 | ||
| GenBank mRNA | AB055759.1 | - | 5 | 3448 | ||
| D87257.1 | - | 2 | 2869 | |||