siRNA Details
| Gene Symbol: Erc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170787.2 | NP_740768.1 | 1 | 219 | ||
| GenBank mRNA | AF541925.1 | - | 1 | 219 | ||
| AY049038.1 | - | 1 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erc2| Gene Symbol: Erc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170787.2 | NP_740768.1 | 1 | 219 | ||
| GenBank mRNA | AF541925.1 | - | 1 | 219 | ||
| AY049038.1 | - | 1 | 277 | |||