siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007606.1 | NP_001007607.1 | 11 | 1462 | ||
| XM_006233136.2 | XP_006233198.1 | 12 | 1569 | |||
| GenBank mRNA | BC061765.1 | - | 11 | 1462 | 5620893 | |
| FQ233851.1 | - | 4 | 447 | |||

Sars
Genomic Map