siRNA Details
| Gene Symbol: Brinp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173115.1 | NP_775138.1 | 8 | 2863 | ||
| GenBank mRNA | AB077853.1 | - | 8 | 2863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brinp2| Gene Symbol: Brinp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173115.1 | NP_775138.1 | 8 | 2863 | ||
| GenBank mRNA | AB077853.1 | - | 8 | 2863 | ||