siRNA Details
| Gene Symbol: Fads3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173137.1 | NP_775160.1 | 11 | 1338 | ||
| GenBank mRNA | AJ494720.1 | - | 11 | 1338 | ||
| HQ901598.1 | - | 11 | 1262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fads3| Gene Symbol: Fads3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173137.1 | NP_775160.1 | 11 | 1338 | ||
| GenBank mRNA | AJ494720.1 | - | 11 | 1338 | ||
| HQ901598.1 | - | 11 | 1262 | |||