siRNA Details
| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100647.1 | NP_001094117.1 | 12 | 1290 | ||
| GenBank mRNA | BC160929.1 | - | 12 | 1290 | 8364923 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fxr2| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100647.1 | NP_001094117.1 | 12 | 1290 | ||
| GenBank mRNA | BC160929.1 | - | 12 | 1290 | 8364923 | |