siRNA Details
| Gene Symbol: Olr1466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 396 | ||
| XM_008767806.1 | XP_008766028.1 | 2 | 1714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1466| Gene Symbol: Olr1466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 396 | ||
| XM_008767806.1 | XP_008766028.1 | 2 | 1714 | |||