siRNA Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105839.1 | NP_001099309.1 | 3 | 472 | ||
| XM_006247274.2 | XP_006247336.1 | 3 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkiras2| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105839.1 | NP_001099309.1 | 3 | 472 | ||
| XM_006247274.2 | XP_006247336.1 | 3 | 423 | |||