siRNA Details
| Gene Symbol: Usp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100501.1 | NP_001093971.1 | 8, 9 | 1010 | ||
| XM_006248015.2 | XP_006248077.1 | 8, 9 | 1026 | |||
| GenBank mRNA | BC087705.1 | - | 8, 9 | 953 | ||
| BC112386.1 | - | 8, 9 | 960 | |||
| CV112036.1 | - | 1, 2 | 113 | |||
| FQ221582.1 | - | 8, 9 | 965 | |||

Usp16
Genomic Map