siRNA Details
| Gene Symbol: Rsph10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013867.1 | NP_001013889.1 | 11, 12 | 1599 | ||
| XM_006248858.2 | XP_006248920.1 | 12, 13 | 2426 | |||
| XM_006248859.2 | XP_006248921.1 | 12, 13 | 2294 | |||
| XM_006248860.2 | XP_006248922.1 | 12, 13 | 2426 | |||
| XM_006248861.2 | XP_006248923.1 | 11, 12 | 2335 | |||
| GenBank mRNA | BC081935.1 | - | 11, 12 | 1599 | 7114952 | |
| FQ212099.1 | - | 11, 12 | 1706 | |||

Rsph10b
Genomic Map