siRNA Details
| Gene Symbol: Gpr161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751260.3 | XP_003751308.1 | 3 | 844 | ||
| XM_006250191.2 | XP_006250253.1 | 3 | 914 | |||
| XM_008769710.1 | XP_008767932.1 | 4 | 925 | |||
| XM_008769711.1 | XP_008767933.1 | 3 | 863 | |||
| XM_008769712.1 | XP_008767934.1 | 3 | 842 | |||
More Information
Set Membership: |
|

Gpr161
Genomic Map