siRNA Details
| Gene Symbol: Tlr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145828.1 | NP_001139300.1 | 1 | 1480 | ||
| XM_006250376.1 | XP_006250438.1 | 5 | 1752 | |||
| XM_006250377.2 | XP_006250439.1 | 4 | 2005 | |||
| XM_008769824.1 | XP_008768046.1 | 2 | 1344 | |||
| XM_008769825.1 | XP_008768047.1 | 2 | 3707 | |||
| XM_008769826.1 | XP_008768048.1 | 4 | 3075 | |||
| GenBank mRNA | FJ750588.1 | - | 1 | 513 | ||

Tlr5
Genomic Map