siRNA Details
| Gene Symbol: Dhx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191597.1 | NP_001178526.1 | 2 | 434 | ||
| XM_006251031.2 | XP_006251093.1 | 2 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx15| Gene Symbol: Dhx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191597.1 | NP_001178526.1 | 2 | 434 | ||
| XM_006251031.2 | XP_006251093.1 | 2 | 438 | |||