siRNA Details
| Gene Symbol: LOC103690141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770469.1 | XP_008768691.1 | 1 | 170 | ||
| Gene Symbol: Ccdc85a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191553.1 | NP_001178482.1 | 93 | |||

LOC103690141
Genomic Map