siRNA Details
| Gene Symbol: Adam28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181693.1 | NP_859044.1 | 15 | 1610 | ||
| XM_008770820.1 | XP_008769042.1 | 15 | 1621 | |||
| GenBank mRNA | AY283187.1 | - | 15 | 1610 | ||
| Gene Symbol: LOC102550654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770807.1 | XP_008769029.1 | 4 | 301 | ||

Adam28
Genomic Map