siRNA Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203340.1 | NP_976085.1 | 3 | 213 | ||
| XM_006252261.2 | XP_006252323.1 | 4 | 1023 | |||
| XM_008770825.1 | XP_008769047.1 | 1 | 35 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npm2| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203340.1 | NP_976085.1 | 3 | 213 | ||
| XM_006252261.2 | XP_006252323.1 | 4 | 1023 | |||
| XM_008770825.1 | XP_008769047.1 | 1 | 35 | |||