siRNA Details
| Gene Symbol: Map1s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106070.1 | NP_001099540.1 | 5 | 1167 | ||
| XM_008771085.1 | XP_008769307.1 | 5 | 1506 | |||
| XM_008771086.1 | XP_008769308.1 | 6 | 1252 | |||
| GenBank mRNA | BC169058.1 | - | 5 | 1139 | 7932017 | |
| FQ211658.1 | - | 5 | 1176 | |||

Map1s
Genomic Map