siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169115.1 | NP_001162586.1 | 25 | 3102 | ||
| XM_006253134.2 | XP_006253196.1 | 25 | 3023 | |||
| XM_006253135.2 | XP_006253197.1 | 25 | 2958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc11| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169115.1 | NP_001162586.1 | 25 | 3102 | ||
| XM_006253134.2 | XP_006253196.1 | 25 | 3023 | |||
| XM_006253135.2 | XP_006253197.1 | 25 | 2958 | |||