siRNA Details
| Gene Symbol: Nrsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106109.2 | NP_001099579.2 | 4 | 715 | ||
| GenBank mRNA | BC166881.1 | - | 4 | 715 | 6891333 | |
| BC166919.1 | - | 4 | 715 | 6919003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrsn1| Gene Symbol: Nrsn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106109.2 | NP_001099579.2 | 4 | 715 | ||
| GenBank mRNA | BC166881.1 | - | 4 | 715 | 6891333 | |
| BC166919.1 | - | 4 | 715 | 6919003 | ||