This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD1311805 SIRNA Assays ›
Gene Symbol:
RGD1311805
Gene Name:
similar to RIKEN cDNA 2400010D15
Gene Aliases:
C18orf8
Chromosome Location:
Chr.18: 3597240 - 3617160 on Build Rnor_6.0
UniGene ID:
Rn.35239
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s145959

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1311805
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009638.1 NP_001009638.1 19 1902
GenBank mRNA BC087672.1 - 19 1902 7317049

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029623, AK144623, AF003352, AK159691, AK013979, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-bgnf4:80/100.66.76.30:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline