siRNA Details
| Gene Symbol: Mmaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106174.1 | NP_001099644.1 | 3 | 656 | ||
| XM_006255398.2 | XP_006255460.1 | 2 | 338 | |||
| XM_006255399.2 | XP_006255461.1 | 2 | 2477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmaa| Gene Symbol: Mmaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106174.1 | NP_001099644.1 | 3 | 656 | ||
| XM_006255398.2 | XP_006255460.1 | 2 | 338 | |||
| XM_006255399.2 | XP_006255461.1 | 2 | 2477 | |||