siRNA Details
| Gene Symbol: LOC100909916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255170.1 | XP_006255232.1 | 2 | 284 | ||
| XM_008763843.1 | XP_008762065.1 | 3 | 383 | |||
| Gene Symbol: RGD1564492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761693.1 | XP_008759915.1 | 2 | 272 | ||
| Gene Symbol: Trappc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106193.1 | NP_001099663.1 | 3 | 262 | ||
| GenBank mRNA | BC166907.1 | - | 3 | 248 | 6888592 | |
| BC166917.1 | - | 3 | 248 | 6918252 | ||
| BC168734.1 | - | 3 | 248 | 6889050 | ||

LOC100909916
Genomic Map