siRNA Details
| Gene Symbol: Pak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106238.1 | NP_001099708.1 | 7, 8 | 1615 | ||
| XM_008759119.1 | XP_008757341.1 | 7, 8 | 3665 | |||
| GenBank mRNA | BC168940.1 | - | 7, 8 | 1625 | 8363273 | |

Pak4
Genomic Map