This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TJP1 SIRNA Assays ›
Gene Symbol:
Tjp1
Gene Name:
tight junction protein 1
Gene Aliases:
ZO-1
Chromosome Location:
Chr.1: 126146489 - 126249545 on Build Rnor_6.0
UniGene ID:
Rn.101871
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s146926

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tjp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106266.1 NP_001099736.1 23, 24 4342
XM_006229330.2 XP_006229392.1 24, 25 4485
XM_006229331.2 XP_006229393.1 24, 25 4574
XM_006229332.2 XP_006229394.1 24, 25 4864
XM_006229333.2 XP_006229395.1 24, 25 4384
XM_008759501.1 XP_008757723.1 24, 25 4808
XM_008759502.1 XP_008757724.1 24, 25 4808
XM_008759503.1 XP_008757725.1 23, 24 4568
XM_008759504.1 XP_008757726.1 23, 24 4568

Back To Top

More Information


Panther Classification:

Molecular Function -

tight junction

Gene Ontology Categories:

Function(s) Process(es)

blastocyst formation
sensory perception of sound
response to lipopolysaccharide
response to drug
negative regulation of vascular permeability
response to ethanol
response to magnetism
cellular response to glucose stimulus
establishment of endothelial intestinal barrier
protein binding
protein C-terminus binding
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0