siRNA Details
| Gene Symbol: Olr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000114.1 | NP_001000114.1 | 1 | 520 | ||
| Gene Symbol: LOC684403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770643.1 | XP_008768865.1 | 1 | 520 | ||

Olr11
Genomic Map