siRNA Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012460.1 | NP_001012478.1 | 6 | 565 | ||
| GenBank mRNA | BC089897.1 | - | 6 | 565 | 7381516 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sept1| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012460.1 | NP_001012478.1 | 6 | 565 | ||
| GenBank mRNA | BC089897.1 | - | 6 | 565 | 7381516 | |