This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SCYL1 SIRNA Assays ›
Gene Symbol:
Scyl1
Gene Name:
SCY1-like 1 (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.1: 221115992 - 221129666 on Build Rnor_6.0
UniGene ID:
Rn.143751
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s147825

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scyl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011938.1 NP_001011938.1 2 156
XM_006230749.2 XP_006230811.2 2 218
GenBank mRNA BC087141.1 - 2 156 7127562
FQ217187.1 - 2 206

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
retrograde vesicle-mediated transport, Golgi to ER
protein kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9w28n:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline