siRNA Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008511.2 | NP_001008511.2 | 5 | 824 | ||
| XM_006230755.1 | XP_006230817.1 | 5 | 937 | |||
| GenBank mRNA | AY280663.1 | - | 4 | 718 | ||
| BC104701.1 | - | 5 | 824 | |||

Esrra
Genomic Map