This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FFAR4 SIRNA Assays ›
Gene Symbol:
Ffar4
Gene Name:
free fatty acid receptor 4
Gene Aliases:
Gpr120, O3far1
Chromosome Location:
Chr.1: 256786124 - 256804156 on Build Rnor_6.0
UniGene ID:
Rn.82501
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148234

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ffar4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047088.1 NP_001040553.1 2 715
GenBank mRNA AB207868.1 - 2 715

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
regulation of glucose transport
cellular response to hormone stimulus
negative regulation of apoptotic process
hormone secretion
negative regulation of cytokine secretion
negative regulation of inflammatory response
detection of chemical stimulus involved in sensory perception of taste
positive regulation of ERK1 and ERK2 cascade
response to peptide
G-protein coupled receptor activity
fatty acid binding
taste receptor activity
peptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-j42gp:80/100.66.77.87:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline