This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGD735065 SIRNA Assays ›
Gene Symbol:
RGD735065
Gene Name:
similar to GI:13385412-like protein splice form I
Gene Aliases:
-
Chromosome Location:
Chr.20: 6869753 - 6910025 on Build Rnor_6.0
UniGene ID:
Rn.234381
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148496

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD735065
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199379.2 NP_955411.1 6, 7 845
XM_006256172.2 XP_006256234.1 5, 6 1260
XM_006256173.2 XP_006256235.1 5, 6 870
XM_006256174.2 XP_006256236.1 6, 7 959
XM_006256175.2 XP_006256237.1 6, 7 1093
XM_006256176.2 XP_006256238.1 7, 8 1054
XM_006256177.2 XP_006256239.1 6, 7 1350
XM_006256178.2 XP_006256240.1 7, 8 1445
XM_006256180.2 XP_006256242.1 6, 7 1400
XM_006256181.2 XP_006256243.1 5, 6 1258
XM_006256182.2 XP_006256244.1 5, 6 1254
XM_008772724.1 XP_008770946.1 6, 7 1374
XM_008772725.1 XP_008770947.1 5, 6 1001
GenBank mRNA BC062242.1 - 6, 7 846 6888059
FQ213349.1 - 3, 4 461
FQ226355.1 - 3, 4 461

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030561, BC058575, AK198839, AK030596, BC004004, and others.

Gene Ontology Categories:

Function(s)

wound healing
positive regulation of cell cycle
epithelial cell proliferation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline