This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRPM2 SIRNA Assays ›
Gene Symbol:
Trpm2
Gene Name:
transient receptor potential cation channel, subfamily M, member 2
Gene Aliases:
Trpm2-predicted
Chromosome Location:
Chr.20: 11434043 - 11482880 on Build Rnor_6.0
UniGene ID:
Rn.214495
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148525

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trpm2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011559.1 NP_001011559.1 27 3882
XM_006256248.2 XP_006256310.1 28 4072
XM_006256249.2 XP_006256311.1 28 4069
XM_008772811.1 XP_008771033.1 28 4120
GenBank mRNA AB193179.1 - 27 3910
AY749166.1 - 27 3882

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138301, BC141391, AJ344343, AK036731, AJ878415, and others.

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

manganese ion transport
metabolic process
response to hydroperoxide
manganese ion transmembrane transport
cation channel activity
manganese ion transmembrane transporter activity
ADP-ribose diphosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-npkhf:80/100.66.72.248:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline