This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CENPK SIRNA Assays ›
Gene Symbol:
Cenpk
Gene Name:
centromere protein K
Gene Aliases:
Solt
Chromosome Location:
Chr.2: 34312766 - 34373917 on Build Rnor_6.0
UniGene ID:
Rn.40474
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s148750

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cenpk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106407.1 NP_001099877.1 6 723
XM_006231888.1 XP_006231950.1 6 723
XM_006231889.2 XP_006231951.1 6 444
XM_006231890.2 XP_006231952.1 6 352
XM_008760667.1 XP_008758889.1 6 723

Back To Top

More Information


Gene Ontology Categories:

Function(s)

positive regulation of transcription from RNA polymerase II promoter

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4dwcp:80/100.66.74.115:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline