siRNA Details
| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761263.1 | XP_008759485.1 | 3 | 781 | ||
| XM_227367.5 | XP_227367.5 | 2 | 824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crnn| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761263.1 | XP_008759485.1 | 3 | 781 | ||
| XM_227367.5 | XP_227367.5 | 2 | 824 | |||