siRNA Details
| Gene Symbol: Lcmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011956.1 | NP_001011956.1 | 1 | 1507 | ||
| GenBank mRNA | BC083783.1 | - | 1 | 1507 | 7096502 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcmt2| Gene Symbol: Lcmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011956.1 | NP_001011956.1 | 1 | 1507 | ||
| GenBank mRNA | BC083783.1 | - | 1 | 1507 | 7096502 | |